Learn More
Thermo Scientific™ National Vial Crushers
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
$23,976.73
Specifications
Product Line National
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
0339692
View Documents Promotion Details
Thermo Scientific™
C4040VC
Manual Benchtop Vial Crusher N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
  • Breaks and separates liquid from all 1mL to 100mL vials and ampules
  • Capable of processing approximately 5000 2mL vials per hour
  • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
  • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
Specifications
Specifications
National