Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa
Catalog No. 03342005
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Pre-slit:
No
Closure Size:
8-425
Catalog No. Pre-slit Closure Size
03342005 No 8-425
1 missing translation for 'options'
Catalog No. 03342005 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' C401365A
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
  • Septa and polypropylene caps bonded together for an integral system
  • Eliminates septa push-through
  • Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
Specifications
Description Black; Bonded Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 8-425
Color Black
Material PTFE, Silicone, Polypropylene
Pre-slit No
Septum Bonded Red PTFE/White Silicone, 0.005 in.
Product Line National
Closure Size 8-425