missing translation for 'onlineSavingsMsg'
Learn More

Nateglinide 98.0+%, TCI America™

Catalog No. p-7134025
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0912500MG 500 mg
N09121G 1 g
N09125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0912500MG Supplier TCI America Supplier No. N0912500MG
Only null left

Chemical Identifiers

CAS 105816-04-4
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.429
MDL Number MFCD00875706
InChI Key OELFLUMRDSZNSF-OFLPRAFFSA-N
Synonym nateglinide, starlix, fastic, starsis, senaglinide, trazec, sdz-djn-608, nateglinide inn, unii-41x3pwk4o2, chembl783
PubChem CID 5311309
IUPAC Name (2R)-3-phenyl-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoic acid
SMILES CC(C)C1CCC(CC1)C(=O)NC(CC2=CC=CC=C2)C(=O)O

Specifications

Color White
Quantity 500 mg
Formula Weight 317.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nateglinide
RTECSNumber SQ7318950
TSCA No