Learn More
Whirl-Pak™ Stand-Up Sample Bags
GREENER_CHOICE
Stand-up bags make it very easy to add material to the bag and to remove sample for testing or analysis. The flat bottom ensures stability and the wire tabs hold the top open for easy access.
$383.86 - $770.10
Specifications
Description Whirl-Pak™ Stand-Up Bag
Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
View More Specs
Products 5
Catalog Number Mfr. No. Dimensions (L x W) Thickness (English) Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Thickness (English) Capacity (Metric) Price Quantity & Availability  
0181289
GREENER_CHOICE
View Documents Promotion Details
Whirl-Pak™
B01364
18.5 x 7.5 cm (7.25 x 3 in.) 2.50 mil 118 mL
Pack of 500 Each for $383.86
Case of 12 Pack for $4,606.32
Only null left
0181290
GREENER_CHOICE
View Documents Promotion Details
Whirl-Pak™
B01365
23 x 11.5 cm (9 x 4.5 in.) 3 mil 532 mL
Pack of 500 Each for $562.44
Case of 6 Pack for $3,374.64
Only null left
01812125
GREENER_CHOICE
View Documents Promotion Details
Whirl-Pak™
B01401
23 x 15 cm (9 x 6 in.) 3 mil 710 mL
Pack of 500 Each for $628.64
Case of 6 Pack for $3,771.84
Only null left
01812128
GREENER_CHOICE
View Documents Promotion Details
Whirl-Pak™
B01450
38 x 15 cm (15 x 6 in.) 4 mil 1.24 L
Pack of 250 Each for $674.58
Case of 4 Pack for $2,698.32
Only null left
01812129
GREENER_CHOICE
View Documents Promotion Details
Whirl-Pak™
B01451
38 x 19 cm (15 x 7.5 in.) 4 mil 2 L
Pack of 250 Each for $770.10
Case of 2 Pack for $1,540.20
Only null left
Description
Description
Stand-up bags make it very easy to add material to the bag and to remove sample for testing or analysis. The flat bottom ensures stability and the wire tabs hold the top open for easy access.
  • Paper-like finish virtually eliminates ink smearing
  • Surface that will accept a variety of writing instruments
  • Sterilized after manufacturing
Specifications
Specifications
Whirl-Pak™ Stand-Up Bag
Tape and Wire
Single-use
Sterile
Cardboard
Ethylene Oxide Gas
-196°C to +82°C
Not Autoclavable
Clear with White Block
Polyethylene
Yes
Plastic
-320°F to +180°F
Stand-up Bag