missing translation for 'onlineSavingsMsg'
Learn More

Naringin Hydrate 95.0+%, TCI America™

Catalog No. N007325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N007325G Supplier TCI America Supplier No. N007325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10236-47-2
Molecular Formula C27H32O14
Molecular Weight (g/mol) 580.54
MDL Number MFCD00148888,MFCD00149445,MFCD01461988
InChI Key DFPMSGMNTNDNHN-ZPHOTFPESA-N
Synonym Naringenin 7-Rhamnoglucoside
PubChem CID 74787988
IUPAC Name (2S)-7-{[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 25 g
Formula Weight 580.54
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Naringin Hydrate
RTECSNumber QN6340000
TSCA Yes