missing translation for 'onlineSavingsMsg'
Learn More

Naringenin 93.0+%, TCI America™

Catalog No. p-7133600
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00725G 5 g
N007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N00725G Supplier TCI America Supplier No. N00725G

Chemical Identifiers

CAS 67604-48-2
Molecular Formula C15H12O5
Molecular Weight (g/mol) 272.256
MDL Number MFCD00006844
InChI Key FTVWIRXFELQLPI-UHFFFAOYSA-N
Synonym naringenin, 5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, naringenine, 4',5,7-trihydroxyflavanone, +/--naringenin, naringetol, salipurpol, --naringenin, narigenin, salipurol
PubChem CID 932
ChEBI CHEBI:50202
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O

Specifications

Quantity 5 g
Formula Weight 272.26
Percent Purity ≥93.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Naringenin