missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naproxen sodium, 98%
Description
A cyclooxygenase inhibitorNaproxen sodium acts as a nonselective inhibitor of Cox-1 and Cox-2. It serves as an anti-inflammatory agent with analgesic and antipyretic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Naproxen sodium is the sodium salt of naproxen
- Naproxen sodium competitively suppresses prostaglandin formation and pyrogen release by blocking the conversion of arachidonic acid
Application
- It is suitable for cell culture research.
Specifications
Specifications
| Melting Point | 259°C to 262°C |
| Color | White |
| Quantity | 100 g |
| Merck Index | 14,6417 |
| Solubility Information | Soluble in water up to 100mg/mlSoluble in methanol and chloroform; Slightly soluble in ether; Insoluble in water. |
| Formula Weight | 252.24 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Naproxen sodium |
Safety and Handling
| missing translation for 'healthHazard1' | H302-H360 |
| missing translation for 'healthHazard3' | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| EINECSNumber | 247-486-2 |
| RTECSNumber | QJ1047000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only