missing translation for 'onlineSavingsMsg'
Learn More

Naproxen sodium, 98%

Catalog No. AAJ6310306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6310306 5 g
AAJ6310314 25 g
AAJ6310322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6310306 Supplier Thermo Scientific Chemicals Supplier No. J6310306

Naproxen sodium (CAS # 26159-34-2) is an arylacetic acid of the non-steroidal anti-inflammatory drug class. | CAS: 26159-34-2 | C14H13NaO3 | 252.25 g/mol

A cyclooxygenase inhibitorNaproxen sodium acts as a nonselective inhibitor of Cox-1 and Cox-2. It serves as an anti-inflammatory agent with analgesic and antipyretic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Naproxen sodium is the sodium salt of naproxen
  • Naproxen sodium competitively suppresses prostaglandin formation and pyrogen release by blocking the conversion of arachidonic acid

Application

  • It is suitable for cell culture research.

Chemical Identifiers

CAS 26159-34-2
Molecular Formula C14H13NaO3
Molecular Weight (g/mol) 252.25
MDL Number MFCD00058507
InChI Key CDBRNDSHEYLDJV-FVGYRXGTSA-M
Synonym naproxen sodium, anaprox, miranax, naprelan, naproxen sodium salt, anapran, anaprotab, aprowell, floginex, flogogin
PubChem CID 23681059
ChEBI CHEBI:7477
IUPAC Name sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
SMILES [Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O

Specifications

Melting Point 259°C to 262°C
Color White
Quantity 5 g
Merck Index 14,6417
Solubility Information Soluble in water up to 100mg/mlSoluble in methanol and chloroform; Slightly soluble in ether; Insoluble in water.
Formula Weight 252.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Naproxen sodium
Hazard Category H302-H360
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 247-486-2
RTECSNumber QJ1047000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only