missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naproxen, 99.3%, MP Biomedicals™
$312.03 - $312.03
Chemical Identifiers
| CAS | 22204-53-1 |
|---|---|
| Molecular Formula | C14H14O3 |
| Molecular Weight (g/mol) | 230.263 |
| InChI Key | CMWTZPSULFXXJA-VIFPVBQESA-N |
| Synonym | naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno |
| PubChem CID | 156391 |
| ChEBI | CHEBI:7476 |
| IUPAC Name | (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid |
| SMILES | CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19024725
|
MP Biomedicals Inc
0219024725 |
25 g |
Each for $312.03
|
|
|||||
Chemical Identifiers
| 22204-53-1 | |
| 230.263 | |
| naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno | |
| CHEBI:7476 | |
| CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O |
| C14H14O3 | |
| CMWTZPSULFXXJA-VIFPVBQESA-N | |
| 156391 | |
| (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid |
Specifications
| 22204-53-1 | |
| 0.993 | |
| 25 g | |
| CMWTZPSULFXXJA-VIFPVBQESA-N | |
| (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid | |
| 156391 | |
| 230.3 | |
| Naproxen |
| White | |
| C14H14O3 | |
| naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno | |
| CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O | |
| 230.263 | |
| CHEBI:7476 | |
| Powder |