missing translation for 'onlineSavingsMsg'
Learn More

α-Naphthyl phosphate, 99.8%, MP Biomedicals™

Catalog No. p-4403672
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10025701 1 g
ICN10025705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10025701 Supplier MP Biomedicals Inc Supplier No. 0210025701
Only null left

Chemical Identifiers

CAS 2183-17-7
Molecular Formula C10H7Na2O4P
Molecular Weight (g/mol) 268.12
MDL Number MFCD00041007
InChI Key QYURIFWAOPAPAJ-UHFFFAOYSA-L
Synonym disodium 1-naphthyl phosphate, sodium naphthalen-1-yl phosphate, disodium naphthyl phosphate, 1-naphthyl phsophoric acid sodium salt, disodium naphthalen-1-yl phosphate, alpha-naphthyl phosphate disodium salt, dipotassium naphthalen-1-yl phosphate, disodium 1-naphthyl phosphate hydrate, sodium alpha-naphthyl phosphate, 1-naphthyl disodium orthophosphate
PubChem CID 75132
IUPAC Name disodium naphthalen-1-yl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Color White
Assay Percent Range 0.998
Quantity 1 g
Formula Weight 268.1
Physical Form Powder
Chemical Name or Material α-Naphthyl phosphate