missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Naphthol Green B

Catalog No. AAA1826814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1826814 25 g
AAA1826822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1826814 Supplier Thermo Scientific Chemicals Supplier No. A1826814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol

It finds its applications as a leather and fur dye and as a biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its applications as a leather and fur dye and as a biological stain.

Solubility
Soluble in water (160 mg/ml at 20°C), ethanol (0.9 mg/ml), and ethylene (9 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 19381-50-1
Molecular Formula C30H18FeN3Na3O15S3
Molecular Weight (g/mol) 881.474
MDL Number MFCD00003886
InChI Key VLBLPLXOYXEXJK-UHFFFAOYSA-K
Synonym Acid Green 1; C.I. 10020
PubChem CID 14598749
IUPAC Name trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron
SMILES C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]

Specifications

Solubility Information Soluble in water (160mg/ml at 20°C),ethanol (0.9mg/ml),and ethylene (9mg/ml).
Formula Weight 878.47
Quantity 25 g
Chemical Name or Material Naphthol Green B
EINECSNumber 243-010-2
RTECSNumber LJ8930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only