missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Naphthenic acid

Catalog No. 703401L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
703401L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 703401L Supplier Sigma Aldrich Supplier No. 703401L*
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Naphthenic acid is commonly used in the synthesis of useful metal naphthenates such as copper naphthenate, a wood preservative; titanium naphthenate, a precursor for the preparation of titanium oxide thin films and a rare earth naphthenate, a lubricant oil additive
  • It can also be in the synthesis of biodegradable naphthenic acid ionic liquids

Specifications

CAS 1338-24-5
Density 0.92 g/mL (at 20°C (literature))
Refractive Index n20/D 1.45
MDL Number MFCD00147696
Quantity 1 L