missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene Zone Refined (number of passes:23) 99.8+%, TCI America™

Supplier:  TCI America N00041SAMPLE

Catalog No. N0004ZONE


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Naphthalene Zone Refined (number of passes:23)
81°C
218°C
MFCD00001742
1334
UFWIBTONFRDIAS-UHFFFAOYSA-N
naphthalene
931
128.17
Crystalline Powder
91-20-3
White
C10H8
ZONE
naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
C1=CC2=CC=CC=C2C=C1
128.17
CHEBI:16482
≥99.8% (GC)

Chemical Identifiers

91-20-3
128.17
UFWIBTONFRDIAS-UHFFFAOYSA-N
931
naphthalene
C10H8
MFCD00001742
naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
CHEBI:16482
C1=CC2=CC=CC=C2C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0311

RTECSNumber : QJ0525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions