missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 220°C/428°F |
| Packaging | Poly Pail |
| Quantity | 50 kg |
| Percent Purity | 0.99 |
| Grade | Ungraded |
| Physical Form | Crystals or Flakes |
| Chemical Name or Material | Naphthalene |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.