missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-boronic acid pinacol ester, 97%

Catalog No. p-7055951
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6467103 1 g
AAH6467106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6467103 Supplier Thermo Scientific Chemicals Supplier No. H6467103

CAS: 256652-04-7 | C16H19BO2 | 254.136 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 256652-04-7
Molecular Formula C16H19BO2
Molecular Weight (g/mol) 254.136
MDL Number MFCD05663868
InChI Key SPPZBAGKKBHZRW-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane, naphthalene-2-boronic acid pinacol ester, naphthalene-2-boronic acid, pinacol ester, 2-naphthaleneboronic acid, pinacol ester, 2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl, 2-naphthaleneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane, amtb668
PubChem CID 10753409
IUPAC Name 4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 254.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Naphthalene-2-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only