missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-carbonitrile, 95%

CAS: 86-53-3 | C11H7N | 153.184 g/mol

$48.90 - $166.30

Chemical Identifiers

CAS 86-53-3
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00003866
InChI Key YJMNOKOLADGBKA-UHFFFAOYSA-N
Synonym 1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
PubChem CID 6846
IUPAC Name naphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1449806
View Documents
Thermo Scientific Chemicals
L1449806
5 g
Each for $48.90
Only null left
 
AAL1449814
View Documents
Thermo Scientific Chemicals
L1449814
25 g
Each for $166.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86-53-3
153.184
YJMNOKOLADGBKA-UHFFFAOYSA-N
6846
C1=CC=C2C(=C1)C=CC=C2C#N
C11H7N
MFCD00003866
1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
naphthalene-1-carbonitrile

Specifications

86-53-3
299°C
MFCD00003866
UN3439
1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
C1=CC=C2C(=C1)C=CC=C2C#N
153.184
153.18
Naphthalene-1-carbonitrile
33°C to 38°C
C11H7N
5 g
971255
YJMNOKOLADGBKA-UHFFFAOYSA-N
naphthalene-1-carbonitrile
6846
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 201-679-8

RTECSNumber : QL5976300

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only