missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-carbonitrile, 95%

Catalog No. AAL1449806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1449806 5 g
AAL1449814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1449806 Supplier Thermo Scientific Chemicals Supplier No. L1449806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-53-3 | C11H7N | 153.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-53-3
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00003866
InChI Key YJMNOKOLADGBKA-UHFFFAOYSA-N
Synonym 1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
PubChem CID 6846
IUPAC Name naphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2C#N

Specifications

Melting Point 33°C to 38°C
Boiling Point 299°C
Quantity 5 g
UN Number UN3439
Beilstein 971255
Formula Weight 153.18
Percent Purity 95%
Chemical Name or Material Naphthalene-1-carbonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 201-679-8
RTECSNumber QL5976300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only