missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4-dicarbonitrile, 98+%

Catalog No. AAL2024806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2024806 5 g
AAL2024814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2024806 Supplier Thermo Scientific Chemicals Supplier No. L2024806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3029-30-9 | C12H6N2 | 178.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3029-30-9
Molecular Formula C12H6N2
Molecular Weight (g/mol) 178.194
MDL Number MFCD03093626
InChI Key BENSWQOUPJQWMU-UHFFFAOYSA-N
Synonym 1,4-dicyanonaphthalene, 1,4-naphthalenedicarbonitrile, 1,4-dicyanonahthalene, acmc-209heb, ghl.pd_mitscher_leg0.246
PubChem CID 76414
IUPAC Name naphthalene-1,4-dicarbonitrile
SMILES C1=CC=C2C(=CC=C(C2=C1)C#N)C#N

Specifications

Melting Point 208°C to 212°C
Odor Odorless
Quantity 5 g
Beilstein 1870226
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material Naphthalene-1,4-dicarbonitrile
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280f
Wear protective clothing.
EINECSNumber 221-197-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only