missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4,5,8-tetracarboxylic Dianhydride (purified by sublimation) 98.0+%, TCI America™

Catalog No. p-7133957
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07551G 1 g
N07555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07551G Supplier TCI America Supplier No. N07551G
Only null left

Chemical Identifiers

CAS 81-30-1
Molecular Formula C14H4O6
Molecular Weight (g/mol) 268.18
MDL Number MFCD00006915
InChI Key YTVNOVQHSGMMOV-UHFFFAOYSA-N
Synonym 1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride
PubChem CID 6678
SMILES C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O

Specifications

Quantity 1 g
Formula Weight 268.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Naphthalene-1,4,5,8-tetracarboxylic Dianhydride (purified by sublimation)
RTECSNumber QK3695000
TSCA Yes