missing translation for 'onlineSavingsMsg'
Learn More

Naphtha, VM and P, Spectrum™ Chemical

Catalog No. 18604744
Change view
Click to view available options
Quantity:
20 L
4 x 4 L
200 L
4 x 20 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604744 4 x 4 L Steel Can
18604741 20 L Steel Can
18604742 4 x 20 L Steel Can
18604745 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604744 Supplier Spectrum Chemical Mfg Cor Supplier No. N10024LTGLCS4

N1002, 8030-30-6

Spectrum™ Chemical Naphtha, VM and P

Chemical Identifiers

CAS 8030-30-6

Specifications

CAS Max % 1
Quantity 4 x 4 L
Packaging Steel Can
UN Number UN1268
Boiling Point 118°C to 150°C
Chemical Name or Material Naphtha