Learn More
Naphazoline hydrochloride, 99%
CAS: 550-99-2 | C14H14N2·ClH | 246.74 g/mol
$118.92 - $118.92
Chemical Identifiers
| CAS | 550-99-2 |
|---|---|
| Molecular Formula | C14H14N2·ClH |
| Molecular Weight (g/mol) | 246.74 |
| MDL Number | MFCD00012554 |
| InChI Key | DJDFFEBSKJCGHC-UHFFFAOYSA-N |
| Synonym | naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon |
| PubChem CID | 11079 |
| ChEBI | CHEBI:7470 |
| IUPAC Name | 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride |
| SMILES | C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC179660250
|
Thermo Scientific Chemicals
179660250 |
25 g | Plastic bottle |
Each for $118.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 550-99-2 | |
| 246.74 | |
| DJDFFEBSKJCGHC-UHFFFAOYSA-N | |
| 11079 | |
| 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride |
| C14H14N2·ClH | |
| MFCD00012554 | |
| naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon | |
| CHEBI:7470 | |
| C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl |
Specifications
| 550-99-2 | |
| 100.0 | |
| White | |
| 99% | |
| C14H14N2·ClH | |
| 25 g | |
| naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon | |
| DJDFFEBSKJCGHC-UHFFFAOYSA-N | |
| 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride | |
| 11079 | |
| 246.74 | |
| Crystalline Powder |
| 98.5 | |
| 254.0°C to 260.0°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00012554 | |
| 15, 6453 | |
| Solubility in water: 170g/L (20°C). | |
| C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl | |
| 246.74 | |
| CHEBI:7470 | |
| 99% | |
| Naphazoline hydrochloride, 99% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Do not eat,drink or smoke when using this product.
Obtain special instructions before use.
GHS Signal Word: Danger
EINECSNumber : 208-989-2
RTECSNumber : NJ4375000
TSCA : TSCA
RUO – Research Use Only