Learn More
Global Life Sciences NANOSEP CENT 10K, BLUE 24PK DFS Item
Supplier:  Cytiva OD010C33
The 10K Nanosep® Centrifugal Devices with Omega™ Membrane are ideal for the quick and dependable concentration and removal of salt from 50 to 500 µL samples. Usual recoveries have a high percentage of greater than 90%. This product is made of low-binding polypropylene and is compatible to use with standard centrifuge rotors that accept 1.5 mL tubes. It is sold in a pack of 24 in blue.
Catalog No. NC9180511
Only null left
Explore available promotions