missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ NanoDrop UltraC and Qubit 4 Bundle
Description
Accuracy and Sensitivity:
- NanoDrop UltraC: Provides quick and easy quantification of nucleic acids and proteins directly from a small sample volume without the need for reagents. However, it may be less sensitive for low-concentration samples.
- Qubit 4: Uses fluorescent dyes specific to the target molecule, offering high sensitivity and accuracy for low-concentration samples.
Complementary Data:
- NanoDrop UltraC: Offers absorbance-based quantification, which is useful for a broad range of concentrations but in most cases requires the sample to be purified. Acclaro contamination alerts provide the user with information on possible contaminants in the sample.
- Qubit 4: Provides fluorescence-based quantification, which is less affected by contaminants and more specific for the target molecule.
Verification:
- Using both instruments can help verify results. If the NanoDrop and Qubit measurements agree, it increases confidence in the data.
- Discrepancies might indicate the presence of contaminants or other issues that need to be addressed.
Efficiency:
- Combining both tools allows researchers to quickly screen samples with NanoDrop UltraC and then perform more accurate quantification on selected samples using Qubit 4.
Broader Application:
- NanoDrop UltraC: Suitable for routine measurements and quick checks.
- Qubit 4: Ideal for precise quantification needed in downstream applications like qPCR, sequencing, or other sensitive assays.
Specifications
Specifications
| Detector Type | Digital |
| Display | Color Touchscreen |
| Languages | English |
| Light Source | LED |
| Product Line | NanoDrop Qubit |
| Spectral Bandwidth | ≤1.8 nm |
| Type | NanoDrop UltraC and Qubit 4 Bundle |
| Voltage | 12 VDC |
| Wavelength Range | 190 to 850 nm |