missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific NanoDrop UltraC FL dsDNA BR Fluorescence Assay Bundle
Description
- Avoid costly workflow setbacks by gaining a deeper understanding of sample quality with the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers Fluorometers.
- Features integrated Thermo Scientific™ Acclaro™ Sample Intelligence technology for enhanced measurement accuracy and contaminant detection.
- Quantify and qualify DNA, RNA, and protein samples in seconds using only 1 to 2 μL, with full-spectral data to support informed downstream analysis decisions.
- Includes an additional cuvette position for flexible sample handling and expanded measurement options.
- Each bundle provides a NanoDrop™ Ultra fluorescence-capable instrument (NanoDrop Ultra FL or NanoDrop UltraC FL) with the NanoDrop Ultra dsDNA BR Assay Kit.
- Obtain complete DNA and RNA concentration and quality data from a single platform supporting both absorbance and fluorescence measurement modes.
- Equipped with red and blue solid-state LEDs enabling both micro-volume absorbance and fluorescence measurements.
- High Specificity - Fluorescence detection ensures greater selectivity than absorbance-based methods, maintaining accuracy even with complex or impure samples.
- Sample Prep Calculator - integrated tool calculates required dye and buffer volumes to simplify reagent preparation.
- Fully integrated qPCR recipe calculator - uses preloaded or custom kit libraries to automatically calculate reactions based on sample concentration.
- Auto-Measure and Auto-Blank functions streamline multi-sample processing, instantly measuring when the pedestal arm is lowered; easily toggled ON or OFF from the touchscreen.
- Export data directly to Microsoft OneDrive, Google Drive, or Thermo Fisher Connect for seamless cloud storage and sharing.
- Direct LIMS Integration - built-in Restful API enables secure data transfer directly into your LIMS environment.
- Data-to-printer options include Wi-Fi, Ethernet, or USB connection; print from PC using NanoDrop Ultra PC Control software.
- Customizable export options allow preview and selection of data prior to saving to ensure compatibility with downstream workflows.
- For research use only; not for diagnostic purposes.
Specifications
Specifications
| Detector Type | 2048-element CMOS Linear Image Sensor |
| Display | Color Touchscreen |
| Languages | English, Chinese, French, Japanese, German, Spanish, Polish, Italian, Korean |
| Light Source | Xenon Flash, LED |
| Product Line | NanoDrop |
| Spectral Bandwidth | ≤1.8 nm (FWHM at Hg 254 nm) |
| Type | Spectrophotometers Bundle |
| Voltage | 100/240 VAC |
| Wavelength Range | 190 to 850 nm |
| Wavelength Selection | Automatic |