Learn More
Thermo Scientific™ NanoDrop Ultra IQ/OQ Kits
Description
Purchase a Thermo Scientific™ NanoDrop™ Ultra IQ/OQ Kit to receive all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ). Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer and Fluorometer Validation Materials include:
- Information on the validation process
- Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
- Installation and instrument certification
NanoDrop Ultra IQOQ Kit
Includes:
- USB flash drive containing IQ/OQ procedure for NanoDrop Ultra
- CHEM-PR1-KIT
- CHEM-PV-1
NanoDrop UltraC IQOQ Kit
Includes:
- USB flash drive containing IQ/OQ procedure for NanoDrop Ultra
- CHEM-PR1-KIT
- CHEM-PV-1
- Cuvette Qualification Kit (840-257300)
Proper application of these materials ensures your instrument is installed correctly and working within manufacturer's specifications.
Specifications
Specifications
| Product Line | NanoDrop |
| Type | NanoDrop Ultra and IQ/OQ Kit |
| For Use With (Equipment) | NanoDrop Ultra Spectrophotometers and Fluorometers |
| Includes | Documentation and materials to facilitate installation and operational qualification procedures (IQ/OQ) for use with NanoDrop Ultra Spectrophotometers and Fluorometers |