missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific NanoDrop Ultra FL dsDNA & RNA BR Fluorescence Assay Bundle
Description
- Avoid costly setbacks in your workflow with an understanding of sample quality like never before using the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers Fluorometers.
- Innovative Thermo Scientific™ Acclaro™ Sample Intelligence technology is built into every instrument for improved measurement accuracy and contaminant identification.
- Quantify and qualify DNA, RNA, and protein samples in seconds with only 1-2 μL, and obtain full-spectral data before you decide to use samples in downstream applications.
- Additional cuvette position also available for additional flexibility.
- Each bundle includes a NanoDrop™ Ultra fluorescence-capable instrument (NanoDrop Ultra FL or NanoDrop UltraC FL) with both the NanoDrop Ultra dsDNA BR and RNA BR Assay Kits.
- Obtain the most complete information about the concentration and quality of your DNA or RNA samples by making absorbance or fluorescence measurements on the same instrument.
- The NanoDrop Ultra FL and NanoDrop UltraC FL are capable of both micro-volume absorbance and fluorescence measurements, thanks to the inclusion of red and blue solid-state light-emitting diodes (LEDs).
- High Specificity - Fluorescence offers greater selectivity than absorbance-based quantification, enabling accurate detection even in impure or complex samples.
- Sample Prep Calculator-integrated reagent calculator determines amount of dye and buffer needed.
- Fully integrated qPCR recipe calculator - use the built-in library of kits, or add your favorite, to let the software run the math for you. The qPCR recipe calculator suggests how to prepare the reaction based on your sample's concentration.
- Auto-Measure and Auto-Blank functions - multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
- Export to the cloud - export data directly to popular cloud storage services like Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
- Direct LIMS Integration - send data directly into your LIMS with the assistance of an integrated Restful API.
- Easy data-to-printer options - Print your results directly from the NanoDrop Ultra instrument via Wi-Fi, Ethernet or USB printer connection; print exported data from your PC using the Nanodrop Ultra PC Control software.
- Data Your Way - customize and preview the information included in the data file prior to export to ensure it meets any downstream needs.
- For research use only; not for diagnostic purposes.
Specifications
Specifications
| Detector Type | 2048-element CMOS Linear Image Sensor |
| Display | Color Touchscreen |
| Languages | English, Chinese, French, Japanese, German, Spanish, Polish, Italian, Korean |
| Light Source | Xenon Flash, LED |
| Product Line | NanoDrop |
| Spectral Bandwidth | ≤1.8 nm (FWHM at Hg 254 nm) |
| Type | Spectrophotometers Bundle |
| Voltage | 100/240 VAC |
| Wavelength Range | 190 to 850 nm |
| Wavelength Selection | Automatic |