Learn More
Thermo Scientific NanoDrop™ Reagents
Catalog No. CF8
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
CF8 CF-8 Calibration Check Kit
361010886 CF-1 Calibration Check Fluid
361010891 ND PR-1 Reconditioning Kit
13400515 PV-1 Performance Verification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. CF8 Supplier Thermo Scientific Supplier No. CHEMCF8
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with the Thermo Scientific NanoDrop 8000 Spectrophotometer.
  • Ten times more concentrated than other commercially available K2Cr2O7 solutions
  • Packaged in 0.5mL snap-top glass ampules
  • Single-use
Includes:
(2) CF-1 Ampules and 8-well PCR tube strip.
Specifications
Type CF-8 Calibration Check Kit
For Use With (Equipment) NanoDrop 8000 Spectrophotometer
Includes 2 CF-1 Ampules; 8-Well PCR Tube Strip.
Packaging 0.5 mL snap-top glass ampules