Learn More
Thermo Scientific NanoDrop™ Reagents
Certified standards for verifying instrument performance to factory specifications are available separately.
Supplier:  Thermo Scientific CHEMPR1KIT
 View more versions of this product
Catalog No. 361010891
Only null left
Description
Description
Certified standards for verifying instrument performance to factory specifications are available separately.
This kit is useful for reconditioning the sample measurement pedestals to a hydrophobic state if they become “unconditioned” (see user manual for more information). The PR-1 kit consists of a small screw cap vial of specially formulated compound and a supply of 25 convenient applicators. Kit sold separately from instrument.
For use with Thermo Scientific™ NanoDrop™ Spectrophotometers.
Includes:
Small screw cap vial of specially formulated reconditioning compound and 25 convenient swabs
Specifications
Specifications
ND PR-1 Reconditioning Kit
All Thermo Scientific NanoDrop Spectrophotometers
Product Suggestions
Product Suggestions