Learn More
Thermo Scientific NanoDrop™ Reagents
Catalog No. 361010886
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
361010886 CF-1 Calibration Check Fluid
CF8 CF-8 Calibration Check Kit
361010891 ND PR-1 Reconditioning Kit
13400515 PV-1 Performance Verification
13400532 PV-8 Performance Verification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361010886 Supplier Thermo Scientific Supplier No. CHEMCF1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with Thermo Scientific NanoDrop Spectrophotometers.
  • Ten times more concentrated compared to other K2Cr2O7 calibration solutions
  • Packaged in 0.5mL snap-top glass ampules
  • Intended for single use only
Specifications
Type CF-1 Calibration Check Fluid
For Use With (Equipment) NanoDrop 8000, NanoDrop Lite, NanoDrop 2000, NanoDrop 2000c spectrophotometers
Packaging 0.5 mL snap-top glass ampules