missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific NanoDrop™ Reagents

Catalog No. 13400532
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13400532 PV-8 Performance Verification
361010886 CF-1 Calibration Check Fluid
CF8 CF-8 Calibration Check Kit
361010891 ND PR-1 Reconditioning Kit
13400515 PV-1 Performance Verification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13400532 Supplier Thermo Scientific Supplier No. CHEMPV8

Certified standards for verifying instrument performance to factory specifications are available separately.

For use with NanoDrop Eight UV-Vis Spectrophotometer Solution with certified absorbance values. Use to create a record of correct photometric performance.

Contains: Two vials of CHEM-PV-1, two pastuer pipettes, and two 8-well PCR strip tubes.

Intended for single use only.

Specifications

Type PV-8 Performance Verification
For Use With (Equipment) NanoDrop Eight