missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nanodrop™ One Spectrophotometer with WiFi and Qubit™ 4 Fluorometer

Catalog No. 13400525
Change view
Click to view available options
Type:
Spectrophotometer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13400525 Spectrophotometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13400525 Supplier Thermo Scientific™ Supplier No. NDONEWQUBIT4

missing translation for 'validMainframeMsg'

As a bundled offering, the NanoDrop™ One Spectrophotometer and Qubit™ 4 Fluorometer allow you to obtain the most complete information about the concentration and quality of your DNA, RNA, or protein samples.

  • NanoDrop One Microvolume UV-Vis Spectrophotometer
    • Fast and easy sample evaluation of nucleic acids and proteins with pipette, measure, clean workflow and no sample carryover
    • Modern stand-alone design with local control and high-resolution touchscreen interface, which saves bench space
    • Accurate measurements up to 27500 ng/μL (dsDNA) with extended auto-range path length technology
    • No need for cuvettes or consumables needed so you can pipette directly onto pedestal sample retention system
    • Pre-programmed applications designed specifically for life scientists
    • Enhanced connectivity with data transfer via USB, Ethernet, Bluetooth™ and WiFi options; PC software available for data management
  • Qubit 4 Fluorometer
    • Fast and highly accurate quantitation of DNA, RNA, and protein in less than three seconds per sample
    • Measures intact RNA in less than 5 seconds per sample
    • High levels of accuracy using only 1 to 20 μL of sample, even with very dilute samples
    • On-board Reagent Calculator quickly generates Qubit working solution preparation instructions
    • Stores results from up to 1000 samples
    • Large 5.7-inch, state-of-the-art color touchscreen for easy workflow navigation
    • Graphical display indicates when samples are in the extended range or out of range
    • Small footprint saves space on your bench
    • Exports data to a USB drive or directly to your computer via a USB cable
    • Ability to personalize your Qubit fluorometer with the assays you run most, add new assays, or even create your own assays with the MyQubit software and web tool
    • Language of your choice including English, French, Spanish, Italian, German, simplified Chinese, and Japanese
    • Dynamic Range: 5 orders of magnitude
    • Memory: 4 GB USB flash drive
    • Emission filters: green (510 to 580 nm), red (665 to 720 nm)
    • Excitation filters: blue (430 to 495 nm), red (600 to 645 nm)
    • LED light sources: blue (max 470 nm), red (max 635 nm)
    • Photodiode detectors: 300 to 1000 nm range

Specifications

Certifications/Compliance CE and UL/CSA Approved
Compatibility DYMO LabelWriter 450 printer, Bluetooth keyboard, mouse and barcode reader
Concentration dsDNA: Pedestal 27500 ng/μL; BSA (IgG): Pedestal 820 (400) mg/mL
Connections Three USB-A ports, Ethernet, Bluetooth™, and Wi-Fi
Detection Range dsDNA: Pedestal 2 ng/μL, Cuvette 0.2 ng/μL; BSA (IgG): Pedestal 0.06 (0.03) mg/mL, Cuvette 0.006 (0.003) mg/mL
Detector Type 2048-element CMOS Linear Image Sensor
Display 7 in. High Definition Color Display, 1280 x 800 pixels
Dimensions (L x W x H) 20 x 25.4 x 32.3 cm (8 x 10 x 12.7 in.)
Languages English, French, German, Spanish, Polish, Chinese, Japanese, Korean
Light Source Xenon Flash
Product Line NanoDrop One
Spectral Bandwidth ≤1.8 nm (FWHM at Hg 254 nm)
Type Spectrophotometer
Voltage 12 VDC
Wavelength Range 190 to 850 nm
For Use With (Application) Nucleic Acid A260, A260/A280, A260/A230 and Labeled Nucleic Acids; Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Labeled Proteins, OD600, Kinetics, UV-Vis, and Custom Methods
Includes Qubit™ 4 Fluorometer (Q33226)
Keypad Built-in touchscreen, multipoint capacitive touch
Measurement Time 8 seconds
Pathlength 0.030 to 1.0 mm (auto-ranging)
Photometric Accuracy 3% at 0.97 A, 302 nm (Absorbance expressed at Abs/mm at 25°C)
Photometric Range 10 mm equivalent: Pedestal 0 to 550 A, Cuvette 0 to 1.5 A
Power Consumption Operating: 12 to 18 W; Standby: 5 W
Processor Android™
Repeatability 0.002 A (1.0 mm path) or 1%CV (SD of 10 individual measurements at 0.97 A)
Sample Temperature 37°C
Sample Volume (Metric) 1 μL
System Requirements Windows™ 8.1 and 10, 64 bit
Wavelength Accuracy ±1 nm
Show More Show Less