missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop™ One/OneC Productivity Kits

Catalog No. 13400517
Change view
Click to view available options
Type:
NanoDrop One Productivity Kit
NanoDrop OneC Productivity Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13400517 NanoDrop OneC Productivity Kit
13400516 NanoDrop One Productivity Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13400517 Supplier Thermo Scientific™ Supplier No. NDPP1C

The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.

The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.

The NanoDrop One Productivity Kit includes:

  • NanoDrop-branded Finnpipette™ F1 Adjustable-Volume Pipette , 0.2–2 μL
  • Microfiber screen wipe for NanoDrop One Spectrophotometer
  • NanoDrop PR-1 Reconditioning Compound Kit
  • NanoDrop PV-1 Performance Verification Solution
  • I love my NanoDrop sticker

The NanoDrop OneC Productivity Kit includes:

  • NanoDrop-branded Finnpipette™ F1 Adjustable Volume Pipette , 0.2–2μL
  • Microfiber screen wipe for NanoDrop OneC spectrophotometer
  • NanoDrop PR-1 Reconditioning Compound Kit
  • NanoDrop PV-1 Performance Verification Solution
  • 10mm path quartz cuvette
  • Cuvette stir bars, 5x2mm, pack of 10
  • I love my NanoDrop sticker

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type NanoDrop OneC Productivity Kit
For Use With (Equipment) NanoDrop OneC
Includes
  • NanoDrop-branded Finnpipette™ F1 Adjustable Volume Pipette , 0.2–2μL
  • Finntip™ Pipette Tips, 10 μL, sterile, pack of 96
  • Microfiber screen wipe for NanoDrop OneC Spectrophotometer
  • NanoDrop PR-1 Reconditioning Compound Kit
  • NanoDrop PV-1 Performance Verification Solution
  • 10mm path quartz cuvette
  • Cuvette stir bars, 5x2mm, pack of 10
  • “I love my NanoDrop” sticker
Storage Requirements Store at room temperature