Learn More
Thermo Scientific™ NanoDrop Label Printer - DYMO LabelWriter 550 Turbo
DYMO LabelWriter 550 Turbo for use with the Thermo Scientific NanoDrop One/OneC Spectrophotometer (running software version 2.8 or higher).
Supplier:  Thermo Scientific™ DYMOPRNTRND
Catalog No. DYMOPRNTRND
Only null left
Description
Description
  • The DYMO™ LabelWriter™ 550 Turbo connects to your Thermo Scientific™ NanoDrop™ One/OneC Spectrophotometer via the incluced USB cable
  • The DYMO LabelWriter 550 Turbo is compatible with NanoDrop instruments running software version 2.8 or higher) and connects to the instrument via the included USB cable
  • NanoDrop One instrument software is designed to print data and spectra on DYMO 30256 Labels
  • Labels Included: size 2-5/16 x 4 in. (59 x 102 mm)
Specifications
Specifications
NanoDrop
Label Printer
110/240 V
Product Suggestions
Product Suggestions