missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Tosyl-L-arginine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. T03301G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03301G 1 g
T033025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03301G Supplier TCI America Supplier No. T03301G

Chemical Identifiers

CAS 1784-03-8
Molecular Formula C14H23ClN4O4S
Molecular Weight (g/mol) 378.872
MDL Number MFCD00012578
InChI Key JIQFFACVQXXHMY-YDALLXLXSA-N
Synonym tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
PubChem CID 2723792
IUPAC Name methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl

Specifications

Melting Point 147°C
Color White
Quantity 1 g
Formula Weight 378.87
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Tosyl-L-arginine Methyl Ester Hydrochloride