missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine 98.0+%, TCI America™

Catalog No. B22535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22535G 5 g
B225325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22535G Supplier TCI America Supplier No. B22535G
Only null left

Chemical Identifiers

CAS 2480-93-5
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.414
MDL Number MFCD00038259
InChI Key QYYCZJUFHDLLOJ-AWEZNQCLSA-N
Synonym boc-orn z-oh, s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-orn cbz-oh, nalpha-boc-ndelta-cbz-l-ornithine, nalpha-tert-butoxycarbonyl-ndelta-carbobenzoxy-l-ornithine, 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-phenylmethoxycarbonylamino pentanoic acid, 2s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-tert-butoxycarbonyl-n'-benzyloxycarbonyl-l-ornithine, n-alpha-boc-n-delta-z-l-ornithine, bocnh-orn z-oh
PubChem CID 7269338
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 366.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine