missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-lysine 98.0+%, TCI America™

Supplier:  TCI America B16695G

Catalog No. B16695G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nalpha-(tert-Butoxycarbonyl)-L-lysine
White-Yellow
MFCD00038203
DQUHYEDEGRNAFO-SVGMAFHSNA-N
(2S)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
246.31
246.31
Crystal/Lump
13734-28-6
C11H22N2O4
boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine
CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O
5 g
2733284
≥98.0% (T)

Chemical Identifiers

13734-28-6
246.31
DQUHYEDEGRNAFO-SVGMAFHSNA-N
2733284
CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O
C11H22N2O4
MFCD00038203
boc-lys-oh, boc-lysine, n-boc-l-lysine, boc-l-lysine, s-6-amino-2-tert-butoxycarbonyl amino hexanoic acid, nalpha-tert-butoxycarbonyl-l-lysine, n-alpha-tert-butoxycarbonyl-l-lysine, nalpha-boc-l-lysine, boc-lys, n alpha-boc-l-lysine
(2S)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions