missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-histidine 98.0+%, TCI America™

Catalog No. p-7119173
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16345G 5 g
B163425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16345G Supplier TCI America Supplier No. B16345G
Only null left

Chemical Identifiers

CAS 17791-52-5
Molecular Formula C11H17N3O4
Molecular Weight (g/mol) 255.274
MDL Number MFCD00065576
InChI Key AYMLQYFMYHISQO-QMMMGPOBSA-N
Synonym boc-his-oh, n-boc-l-histidine, boc-l-histidine, tert-butyloxycarbonyl-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, boc-l-his-oh, n-tert-butoxycarbonyl-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl
PubChem CID 87308
IUPAC Name (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 255.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-L-histidine