missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-L-asparagine 4-Nitrophenyl Ester 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 146°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 353.33 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalpha-(tert-Butoxycarbonyl)-L-asparagine 4-Nitrophenyl Ester |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |