missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-L-arginine Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. B16265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16265G 5 g
B162625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16265G Supplier TCI America Supplier No. B16265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35897-34-8
Molecular Formula C11H23ClN4O4
Molecular Weight (g/mol) 310.78
MDL Number MFCD00065552
InChI Key HDELGKMVZYHPPB-RZUXQYDTNA-N
Synonym boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o
PubChem CID 2729004
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride
SMILES Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 310.78
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-L-arginine Hydrochloride Monohydrate