missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-L-arginine Hydrochloride Monohydrate 98.0+%, TCI America™
Supplier: TCI America B162625G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 35897-34-8 | |
| 310.78 | |
| HDELGKMVZYHPPB-RZUXQYDTNA-N | |
| 2729004 | |
| Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O |
| C11H23ClN4O4 | |
| MFCD00065552 | |
| boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o | |
| (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride |
Specifications
| Nalpha-(tert-Butoxycarbonyl)-L-arginine Hydrochloride Monohydrate | |
| White | |
| MFCD00065552 | |
| boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o | |
| Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O | |
| 310.78 | |
| 310.78 | |
| Crystalline Powder |
| 35897-34-8 | |
| C11H23ClN4O4 | |
| 25 g | |
| HDELGKMVZYHPPB-RZUXQYDTNA-N | |
| (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride | |
| 2729004 | |
| ≥98.0% (T) |
Safety and Handling
TSCA : No