missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-D-asparagine 98.0+%, TCI America™

Catalog No. p-7119866
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29645G 5 g
B296425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29645G Supplier TCI America Supplier No. B29645G
Only null left

Chemical Identifiers

CAS 75647-01-7
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00065558
InChI Key FYYSQDHBALBGHX-RXMQYKEDSA-N
Synonym boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1623168
IUPAC Name (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 232.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-D-asparagine