missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%

Catalog No. AAJ63959MF
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63959MF 50 mg
AAJ63959MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63959MF Supplier Thermo Scientific Chemicals Supplier No. J63959MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of papain and trypsin | CAS: 4238-41-9 | C14H22Cl2N2O3S | 369.30 g/mol

Inhibitor of papain and trypsin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4238-41-9
Molecular Formula C14H22Cl2N2O3S
Molecular Weight (g/mol) 369.30
MDL Number MFCD00065395
InChI Key YFCUZWYIPBUQBD-ZOWNYOTGSA-N
Synonym 1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride, p-toluenesulfonamide, hydrochloride 7ci, 8ci, nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride
PubChem CID 23335676
IUPAC Name N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride
SMILES Cl.CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl

Specifications

Color White
Solubility Information Soluble in water
Quantity 50 mg
Formula Weight 369.31
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-199-0
RTECSNumber XT5160000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only