missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Ndelta,N^w-Tris(benzyloxycarbonyl)-L-arginine, 95%

Catalog No. AAH6362103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6362103 1 g
AAH6362106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6362103 Supplier Thermo Scientific Chemicals Supplier No. H6362103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14611-34-8 | C30H32N4O8 | 576.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14611-34-8
Molecular Formula C30H32N4O8
Molecular Weight (g/mol) 576.606
MDL Number MFCD00038693
InChI Key YSGAXJCIEJGVFV-RUZDIDTESA-N
Synonym Z-Arg(Z)2-OH
PubChem CID 131843446
IUPAC Name (2R)-2-(phenylmethoxycarbonylamino)-5-[phenylmethoxycarbonyl-[(E)-N'-phenylmethoxycarbonylcarbamimidoyl]amino]pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN(C(=NC(=O)OCC2=CC=CC=C2)N)C(=O)OCC3=CC=CC=C3)C(=O)O

Specifications

Melting Point 139°C
Beilstein 3079013
Quantity 1 g
Formula Weight 576.6
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nalpha,Ndelta,Nomega-Tris(benzyloxycarbonyl)-L-arginine
EINECSNumber 238-647-8
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only