missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%

CAS: 201485-17-8 | C22H25N3O5 | 411.46 g/mol

$124.32 - $477.03

Chemical Identifiers

CAS 201485-17-8
Molecular Formula C22H25N3O5
Molecular Weight (g/mol) 411.46
MDL Number MFCD00237387
InChI Key VJZUCXPETVPQIB-TWYLJJHKNA-N
Synonym fmoc-hocit-oh, fmoc-l-homocitrulline, fmoc-hcit-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid, fmoc-homocit-oh, 2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1383, fmoc-lys carbamoyl-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid
PubChem CID 7019813
IUPAC Name (2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6618903
View Documents
Thermo Scientific Chemicals
H6618903
1 g N/A
Only null left
 
AAH6618906
View Documents
Thermo Scientific Chemicals
H6618906
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

201485-17-8
411.46
VJZUCXPETVPQIB-TWYLJJHKNA-N
7019813
NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C22H25N3O5
MFCD00237387
fmoc-hocit-oh, fmoc-l-homocitrulline, fmoc-hcit-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid, fmoc-homocit-oh, 2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1383, fmoc-lys carbamoyl-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid
(2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid

Specifications

201485-17-8
MFCD00237387
VJZUCXPETVPQIB-TWYLJJHKNA-N
(2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
411.46
411.46
Solid
C22H25N3O5
fmoc-hocit-oh, fmoc-l-homocitrulline, fmoc-hcit-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid, fmoc-homocit-oh, 2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1383, fmoc-lys carbamoyl-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid
NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1 g
7019813
95%
Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only