missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-N^e-(4-methyltrityl)-D-lysine, 97%
CAS: 198544-94-4 | C41H40N2O4 | 624.78 g/mol
Supplier: Thermo Scientific Chemicals H6680306
| Quantity | 5 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 198544-94-4 | |
| 624.78 | |
| YPTNAIDIXCOZAJ-KXQOOQHDSA-N | |
| 51340501 | |
| CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C41H40N2O4 | |
| MFCD26793593 | |
| r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine | |
| (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid |
Specifications
| Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-D-lysine | |
| C41H40N2O4 | |
| r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine | |
| CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 624.78 | |
| 51340501 | |
| 98% |
| 198544-94-4 | |
| MFCD26793593 | |
| YPTNAIDIXCOZAJ-KXQOOQHDSA-N | |
| (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid | |
| 5 g | |
| 624.78 | |
| Solid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold