missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-N^e-(4-methyltrityl)-D-lysine, 97%

CAS: 198544-94-4 | C41H40N2O4 | 624.78 g/mol

Supplier:  Thermo Scientific Chemicals H6680303

Quantity 1 g

 View more versions of this product

Catalog No. AAH6680303


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

198544-94-4
624.78
YPTNAIDIXCOZAJ-KXQOOQHDSA-N
51340501
CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
C41H40N2O4
MFCD26793593
r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid

Specifications

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-D-lysine
C41H40N2O4
r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine
CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
624.78
51340501
98%
198544-94-4
MFCD26793593
YPTNAIDIXCOZAJ-KXQOOQHDSA-N
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
1 g
624.78
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions