missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Carbobenzoxy-L-lysine 97.0+%, TCI America™

Catalog No. C17285G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17285G Supplier TCI America Supplier No. C17285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2212-75-1
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00038204
InChI Key OJTJKAUNOLVMDX-LBPRGKRZSA-N
Synonym z-lys-oh, nalpha-cbz-l-lysine, n-alpha-cbz-l-lysine, cbz-lys-oh, n-alpha-carbobenzoxy-l-lysine, s-6-amino-2-benzyloxy carbonyl amino hexanoic acid, z-lysine, cbz-l-lysine, n-alpha-benzyloxycarbonyl-l-lysine, lysine, n2-phenylmethoxy carbonyl
PubChem CID 75172
IUPAC Name (2S)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid
SMILES NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 280.32
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-L-lysine