missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Carbobenzoxy-L-histidine 99.0+%, TCI America™

Catalog No. p-7118393
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02651G 1 g
B02655G 5 g
B026525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B02651G Supplier TCI America Supplier No. B02651G
Only null left

Chemical Identifiers

CAS 14997-58-1
Molecular Formula C14H15N3O4
Molecular Weight (g/mol) 289.291
MDL Number MFCD00065960
InChI Key WCOJOHPAKJFUDF-LBPRGKRZSA-N
Synonym z-his-oh, n-cbz-l-histidine, cbz-his-oh, cbz-l-his-oh, nalpha-cbz-l-histidine, cbz-histidine, nalpha-carbobenzoxy-l-histidine, l-histidine, n-phenylmethoxy carbonyl, z-l-histidine, s-2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid
PubChem CID 736313
IUPAC Name (2S)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O

Specifications

Melting Point 163°C
Color White
Quantity 1 g
Formula Weight 289.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-L-histidine