missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Carbobenzoxy-D-lysine 98.0+%, TCI America™
$83.89 - $249.78
Chemical Identifiers
| CAS | 70671-54-4 |
|---|---|
| Molecular Formula | C14H20N2O4 |
| Molecular Weight (g/mol) | 280.32 |
| MDL Number | MFCD00067713 |
| InChI Key | OJTJKAUNOLVMDX-GFCCVEGCSA-N |
| Synonym | z-d-lys-oh, a-cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, nalpha-carbobenzoxy-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, nalpha-z-d-lysine, n |
| PubChem CID | 7017997 |
| IUPAC Name | (2R)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid |
| SMILES | NCCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| 70671-54-4 | |
| 280.32 | |
| OJTJKAUNOLVMDX-GFCCVEGCSA-N | |
| 7017997 | |
| NCCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
| C14H20N2O4 | |
| MFCD00067713 | |
| z-d-lys-oh, a-cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, nalpha-carbobenzoxy-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, nalpha-z-d-lysine, n | |
| (2R)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid |
Specifications
| 218°C | |
| White | |
| MFCD00067713 | |
| OJTJKAUNOLVMDX-GFCCVEGCSA-N | |
| (2R)-6-amino-2-{[(benzyloxy)carbonyl]amino}hexanoic acid | |
| 280.32 | |
| 280.32 | |
| Crystalline Powder |
| 70671-54-4 | |
| C14H20N2O4 | |
| z-d-lys-oh, a-cbz-d-lysine, cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, n-alpha-benzyloxycarbonyl-d-lysine, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, nalpha-z-d-lysine, n | |
| NCCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 1 g | |
| 7017997 | |
| ≥98.0% (HPLC,T) | |
| Nalpha-Carbobenzoxy-D-lysine |
Safety and Handling
TSCA : No