missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Carbobenzoxy-D-asparagine 99.0+%, TCI America™
$54.06 - $145.40
Chemical Identifiers
| CAS | 4474-86-6 |
|---|---|
| Molecular Formula | C12H14N2O5 |
| Molecular Weight (g/mol) | 266.25 |
| MDL Number | MFCD00065696 |
| InChI Key | FUCKRCGERFLLHP-UHFFFAOYNA-N |
| Synonym | n-cbz-d-asparagine, z-d-asn-oh, nalpha-cbz-d-asparagine, z-d-asparagine, cbz d-asparagine, cbz-d-asn-oh, n-carbobenzoxy-d-asparagine, n-cbz-d-asn-oh, nalpha-carbobenzoxy-d-asparagine, 2r-2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid |
| PubChem CID | 1712147 |
| IUPAC Name | 2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid |
| SMILES | NC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| 4474-86-6 | |
| 266.25 | |
| FUCKRCGERFLLHP-UHFFFAOYNA-N | |
| 1712147 | |
| NC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
| C12H14N2O5 | |
| MFCD00065696 | |
| n-cbz-d-asparagine, z-d-asn-oh, nalpha-cbz-d-asparagine, z-d-asparagine, cbz d-asparagine, cbz-d-asn-oh, n-carbobenzoxy-d-asparagine, n-cbz-d-asn-oh, nalpha-carbobenzoxy-d-asparagine, 2r-2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid | |
| 2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid |
Specifications
| 4474-86-6 | |
| White | |
| MFCD00065696 | |
| n-cbz-d-asparagine, z-d-asn-oh, nalpha-cbz-d-asparagine, z-d-asparagine, cbz d-asparagine, cbz-d-asn-oh, n-carbobenzoxy-d-asparagine, n-cbz-d-asn-oh, nalpha-carbobenzoxy-d-asparagine, 2r-2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid | |
| NC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 266.25 | |
| 266.25 | |
| Crystalline Powder |
| 165°C | |
| C12H14N2O5 | |
| 100 mg | |
| FUCKRCGERFLLHP-UHFFFAOYNA-N | |
| 2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid | |
| 1712147 | |
| ≥99.0% (T) | |
| Nalpha-Carbobenzoxy-D-asparagine |
Safety and Handling
TSCA : No